2,4-Dichloro-5-methoxy-N-methylaniline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1350-5G AN-1350-1G
COBBAN-1350-5GEA 0 GBP
COBBAN-1350-5G COBBAN-1350-1G
2,4-Dichloro-5-methoxy-N-methylaniline
2,4-Dichloro-5-methoxy-N-methylaniline
Formula: C₈H₉Cl₂NO
MDL Number: MFCD11504829
CAS Number: 1072945-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR