3-Methoxy-5-(methoxycarbonyl)phenylboronic acid pinacol ester
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PN-5505-1G
PN-5505-250M
COBBPN-5505-1GEA
0
GBP
COBBPN-5505-1G
COBBPN-5505-250M
3-Methoxy-5-(methoxycarbonyl)phenylboronic acid pinacol ester
3-Methoxy-5-(methoxycarbonyl)phenylboronic acid pinacol ester
Formula:
C₁₅H₂₁BO₅ |
MDL Number:
MFCD12026074 CAS Number: 889654-06-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...