3-Ethoxy-5-(methoxycarbonyl)phenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PN-5972-1G PN-5972-250M
COBBPN-5972-1GEA 0 GBP
COBBPN-5972-1G COBBPN-5972-250M
3-Ethoxy-5-(methoxycarbonyl)phenylboronic acid pinacol ester
3-Ethoxy-5-(methoxycarbonyl)phenylboronic acid pinacol ester
Formula: C₁₆H₂₃BO₅
MDL Number: MFCD12546593
CAS Number: 1218789-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR