4-(Diethylcarbamoyl)phenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(N,N-Diethylaminocarbonyl)phenylboronic acid, pinacol ester, [4-(Diethylamine-1-carbonyl)phenyl]boronic acid pinacol ester, N,N-Diethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

PN-3001-5G PN-3001-1G
COBBPN-3001-5GEA 0 GBP
COBBPN-3001-5G COBBPN-3001-1G
4-(Diethylcarbamoyl)phenylboronic acid pinacol ester
4-(Diethylcarbamoyl)phenylboronic acid pinacol ester
Formula: C₁₇H₂₆BNO₃
MW: 303.2000 g/mol
MDL Number: MFCD06795673
CAS Number: 325142-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR