1-Methylimidazole-2-carbaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Methyl-1H-imidazole-2-carbaldehyde, 1-Methylimidazole-2-carboxaldehyde

AS32540G1 AS32540G5
ACTIAS32540G1EA 0 GBP
ACTIAS32540G1 ACTIAS32540G5
1-Methylimidazole-2-carbaldehyde
1-Methylimidazole-2-carbaldehyde
Formula: C₅H₆N₂O
MDL Number: MFCD01321308
CAS Number: 13750-81-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR