1-Bromo-1-cycloheptene
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-0604-5G
OT-0604-1G
COBBOT-0604-5GEA
0
GBP
COBBOT-0604-5G
COBBOT-0604-1G
1-Bromo-1-cycloheptene
1-Bromo-1-cycloheptene
Formula:
C₇H₁₁Br MW: 175.066 g/mol |
MDL Number:
MFCD09878419 CAS Number: 18317-64-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...