6-Fluoro-4-quinolinol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF1103.1 FL5483.500 005483-1G 005483-5G
CHMPCFF1103.1EA 0 GBP
CHMPCFF1103.1 CHMPFL5483.500 FLUO005483-1G FLUO005483-5G
6-Fluoro-4-quinolinol
6-Fluoro-4-quinolinol
Formula: C₉H₆FNO
MDL Number: MFCD00269610
CAS Number: 391-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR