6-Fluoro-4-quinolinol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20940G1 AS20940G25 AS20940G5
ACTIAS20940G1EA 0 GBP
ACTIAS20940G1 ACTIAS20940G25 ACTIAS20940G5
6-Fluoro-4-quinolinol
6-Fluoro-4-quinolinol
Formula: C₉H₆FNO
MDL Number: MFCD00269610
CAS Number: 391-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR