2-Bromo-N-(4-methoxybenzyl)-4-nitroaniline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2678-5G
HC-2678-1G
COBBHC-2678-5GEA
0
GBP
COBBHC-2678-5G
COBBHC-2678-1G
2-Bromo-N-(4-methoxybenzyl)-4-nitroaniline
2-Bromo-N-(4-methoxybenzyl)-4-nitroaniline
Formula:
C₁₄H₁₃BrN₂O₃ |
MDL Number:
MFCD12026040 CAS Number: 1150271-16-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...