4-Bromo-N-methyl-2-nitroaniline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AN-1088-5G
AN-1088-1G
COBBAN-1088-5GEA
0
GBP
COBBAN-1088-5G
COBBAN-1088-1G
4-Bromo-N-methyl-2-nitroaniline
4-Bromo-N-methyl-2-nitroaniline
Formula:
C₇H₇BrN₂O₂ MW: 231.0467 g/mol |
MDL Number:
MFCD00624242 CAS Number: 53484-26-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...