4-Bromo-N-ethyl-2-nitroaniline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AN-1265-5G AN-1265-1G
COBBAN-1265-5GEA 0 GBP
COBBAN-1265-5G COBBAN-1265-1G
4-Bromo-N-ethyl-2-nitroaniline
4-Bromo-N-ethyl-2-nitroaniline
Formula: C₈H₉BrN₂O₂
MW: 245.0733 g/mol
MDL Number: MFCD00768959
CAS Number: 56136-82-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR