4-Bromo-N-isopropyl-2-nitroaniline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2735-5G HC-2735-1G
COBBHC-2735-5GEA 0 GBP
COBBHC-2735-5G COBBHC-2735-1G
4-Bromo-N-isopropyl-2-nitroaniline
4-Bromo-N-isopropyl-2-nitroaniline
Formula: C₉H₁₁BrN₂O₂
MDL Number: MFCD03407977
CAS Number: 683274-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR