2-Bromo-N-methyl-4-nitroaniline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2679-5G HC-2679-1G
COBBHC-2679-5GEA 0 GBP
COBBHC-2679-5G COBBHC-2679-1G
2-Bromo-N-methyl-4-nitroaniline
2-Bromo-N-methyl-4-nitroaniline
Formula: C₇H₇BrN₂O₂
MDL Number: MFCD11052559
CAS Number: 6911-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR