2-Bromo-N-ethyl-4-nitroaniline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2681-5G
HC-2681-1G
COBBHC-2681-5GEA
0
GBP
COBBHC-2681-5G
COBBHC-2681-1G
2-Bromo-N-ethyl-4-nitroaniline
2-Bromo-N-ethyl-4-nitroaniline
Formula:
C₈H₉BrN₂O₂ |
MDL Number:
MFCD12044149 CAS Number: 108485-08-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...