Potassium (2-fluoro-3-isopropoxyphenyl)trifluoroborate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

TB-5281-1G TB-5281-5G
COBBTB-5281-1GEA 0 GBP
COBBTB-5281-1G COBBTB-5281-5G
Potassium (2-fluoro-3-isopropoxyphenyl)trifluoroborate
Potassium (2-fluoro-3-isopropoxyphenyl)trifluoroborate
Formula: C₉H₁₀BF₄KO
MDL Number: MFCD11504989
CAS Number: 1073468-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR