2-(Ethoxycarbonyl)phenylboronic acid propanediol cyclic ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 2-(1,3,2-dioxaborinan-2-yl)benzoate

PR-2126-5G PR-2126-1G
COBBPR-2126-5GEA 0 GBP
COBBPR-2126-5G COBBPR-2126-1G
2-(Ethoxycarbonyl)phenylboronic acid propanediol cyclic ester
2-(Ethoxycarbonyl)phenylboronic acid propanediol cyclic ester
Formula: C₁₂H₁₅BO₄
MW: 234.0561 g/mol
MDL Number: MFCD06659932
CAS Number: 850567-60-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR