2,4-Dichloro-6-fluoroquinazoline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS2587G1 AS2587G5
ACTIAS2587G1EA 0 GBP
ACTIAS2587G1 ACTIAS2587G5
2,4-Dichloro-6-fluoroquinazoline
2,4-Dichloro-6-fluoroquinazoline
Formula: C₈H₃Cl₂FN₂
MDL Number: MFCD09954884
CAS Number: 134517-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR