4,7-Dichloroquinazoline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS2552G1 AS2552G25 AS2552G5
ACTIAS2552G1EA 0 GBP
ACTIAS2552G1 ACTIAS2552G25 ACTIAS2552G5
4,7-Dichloroquinazoline
4,7-Dichloroquinazoline
Formula: C₈H₄Cl₂N₂
MW: 199.0368 g/mol
MDL Number: MFCD00023911
CAS Number: 2148-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR