8-Bromo-2,4-dichloroquinazoline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS2585G1 AS2585G5
ACTIAS2585G1EA 0 GBP
ACTIAS2585G1 ACTIAS2585G5
8-Bromo-2,4-dichloroquinazoline
8-Bromo-2,4-dichloroquinazoline
Formula: C₈H₃BrCl₂N₂
MW: 277.93 g/mol
MDL Number: MFCD09954891
CAS Number: 331647-05-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR