7-Bromo-4-chloroquinazoline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS2476G1 AS2476G5
ACTIAS2476G1EA 0 GBP
ACTIAS2476G1 ACTIAS2476G5
7-Bromo-4-chloroquinazoline
7-Bromo-4-chloroquinazoline
Formula: C₈H₄BrClN₂
MDL Number: MFCD06657216
CAS Number: 573675-55-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR