7-Bromo-2,4-dichloroquinazoline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22664G1 AS22664G5
ACTIAS22664G1EA 57 GBP
ACTIAS22664G1 ACTIAS22664G5
7-Bromo-2,4-dichloroquinazoline
7-Bromo-2,4-dichloroquinazoline
Formula: C₈H₃BrCl₂N₂
MW: 277.93 g/mol
MDL Number: MFCD09954890
CAS Number: 959237-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR