6-Bromo-2-chloroquinazoline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS2480G1 AS2480G5
ACTIAS2480G1EA 0 GBP
ACTIAS2480G1 ACTIAS2480G5
6-Bromo-2-chloroquinazoline
6-Bromo-2-chloroquinazoline
Formula: C₈H₄BrClN₂
MDL Number: MFCD09261000
CAS Number: 882672-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR