6-Bromo-2,4-dichloroquinazoline
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS2291G1
AS2291G5
ACTIAS2291G1EA
0
GBP
ACTIAS2291G1
ACTIAS2291G5
6-Bromo-2,4-dichloroquinazoline
6-Bromo-2,4-dichloroquinazoline
Formula:
C₈H₃BrCl₂N₂ MW: 277.93 g/mol |
MDL Number:
MFCD09744007 CAS Number: 102393-82-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...