2-(Benzyloxy)-5-bromoanisole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0647-5G OT-0647-1G
COBBOT-0647-5GEA 0 GBP
COBBOT-0647-5G COBBOT-0647-1G
2-(Benzyloxy)-5-bromoanisole
2-(Benzyloxy)-5-bromoanisole
Formula: C₁₄H₁₃BrO₂
MDL Number: MFCD00092521
CAS Number: 63057-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR