3-(Chloromethyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-2478-5G BB-2478-25G BB-2478-1G
COBBBB-2478-5GEA 0 GBP
COBBBB-2478-5G COBBBB-2478-25G COBBBB-2478-1G
3-(Chloromethyl)phenylboronic acid
3-(Chloromethyl)phenylboronic acid
Formula: C₇H₈BClO₂
MDL Number: MFCD09475872
CAS Number: 957035-15-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR