2,6-Dichloro-3-methylphenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BB-5010-5G
BB-5010-25G
BB-5010-1G
COBBBB-5010-5GEA
0
GBP
COBBBB-5010-5G
COBBBB-5010-25G
COBBBB-5010-1G
2,6-Dichloro-3-methylphenylboronic acid
2,6-Dichloro-3-methylphenylboronic acid
Formula:
C₇H₇BCl₂O₂ |
MDL Number:
MFCD09800875 CAS Number: 851756-54-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...