(2-Bromophenyl)acetamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4689-5G CA-4689-1G
COBBCA-4689-5GEA 0 GBP
COBBCA-4689-5G COBBCA-4689-1G
(2-Bromophenyl)acetamide
(2-Bromophenyl)acetamide
Formula: C₈H₈BrNO
MDL Number: MFCD00099254
CAS Number: 65999-53-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR