Ethyl-6-bromopicolinate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Ethyl 6-bromopyridine-2-carboxylate

AS21518G100 AS21518G25
ACTIAS21518G100EA 230 GBP
ACTIAS21518G100 ACTIAS21518G25
Ethyl-6-bromopicolinate
Ethyl 6-bromopicolinate
Formula: C₈H₈BrNO₂
MW: 230.0600 g/mol
MDL Number: MFCD04116913
CAS Number: 21190-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR