Methyl-2-chloro-6-methylisonicotinate
Supplier: FLUOROCHEM LTD (CHEMPUR)
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Methyl 2-chloro-6-picoline-4-carboxylate
, Methyl 2-chloro-6-methylpyridine-4-carboxylate
FLUO019484-2.5GEA
0
GBP
FLUO019484.1G
FLUO019484.5G
FLUO019484-2.5G
Methyl-2-chloro-6-methylisonicotinate
Methyl 2-chloro-6-methylisonicotinate
Formula:
C₈H₈ClNO₂ MW: 185.61 g/mol |
MDL Number:
MFCD04038391 CAS Number: 3998-90-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...