4-(tert-Butylcarbamoyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(tert-Butylcarbamoyl)benzeneboronic acid, (4-(tert-Butylcarbamoyl)phenyl)boronic acid, 4-(tert-Butylaminocarbonyl)phenylboronic acid

BB-3012-5G BB-3012-25G BB-3012-1G
COBBBB-3012-5GEA 0 GBP
COBBBB-3012-5G COBBBB-3012-25G COBBBB-3012-1G
4-(tert-Butylcarbamoyl)phenylboronic acid
4-(tert-Butylcarbamoyl)phenylboronic acid
Formula: C₁₁H₁₆BNO₃
MW: 221.06 g/mol
MDL Number: MFCD03788413
CAS Number: 850568-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR