3-(Butylcarbamoyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(Butylaminocarbonyl)phenylboronic acid, 3-(n-Butylcarbamoyl)benzeneboronic acid, (3-(Butylcarbamoyl)phenyl)boronic acid

BB-3060-5G BB-3060-1G
COBBBB-3060-5GEA 0 GBP
COBBBB-3060-5G COBBBB-3060-1G
3-(Butylcarbamoyl)phenylboronic acid
3-(Butylcarbamoyl)phenylboronic acid
Formula: C₁₁H₁₆BNO₃
MW: 221.06 g/mol
MDL Number: MFCD04115691
CAS Number: 397843-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR