4-(Dimethylcarbamoyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (4-(Dimethylcarbamoyl)phenyl)boronic acid, 4-(Dimethylcarbamoyl)benzeneboronic acid, 4-(N,N-Dimethylaminocarbonyl)phenylboronic acid

BB-3000-5G BB-3000-25G BB-3000-1G
COBBBB-3000-5GEA 0 GBP
COBBBB-3000-5G COBBBB-3000-25G COBBBB-3000-1G
4-(Dimethylcarbamoyl)phenylboronic acid
4-(Dimethylcarbamoyl)phenylboronic acid
Formula: C₉H₁₂BNO₃
MW: 193.01 g/mol
MDL Number: MFCD02258943
CAS Number: 405520-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR