4-(Furfurylcarbamoyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (4-((Furan-2-ylmethyl)carbamoyl)phenyl)boronic acid, 4-[(Fur-2-ylmethyl)carbamoyl]benzeneboronic acid

BB-3019-5G BB-3019-1G
COBBBB-3019-5GEA 0 GBP
COBBBB-3019-5G COBBBB-3019-1G
4-(Furfurylcarbamoyl)phenylboronic acid
4-(Furfurylcarbamoyl)phenylboronic acid
Formula: C₁₂H₁₂BNO₄
MW: 245.0400 g/mol
MDL Number: MFCD04115672
CAS Number: 850568-18-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR