3-(Methylsulphonyl)benzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20734G25 AS20734G5
ACTIAS20734G25EA 0 GBP
ACTIAS20734G25 ACTIAS20734G5
3-(Methylsulphonyl)benzoic acid
3-(Methylsulphonyl)benzoic acid
Formula: C₈H₈O₄S
MW: 200.21 g/mol
MDL Number: MFCD00216488
CAS Number: 5345-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR