4'-Piperidinoacetophenone
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OT-0407-5G
OT-0407-25G
OT-0407-100G
COBBOT-0407-5GEA
0
GBP
COBBOT-0407-5G
COBBOT-0407-25G
COBBOT-0407-100G
4'-Piperidinoacetophenone
4'-Piperidinoacetophenone
Formula:
C₁₃H₁₇NO |
MDL Number:
MFCD00006481 CAS Number: 10342-85-5 EINECS: 233-746-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...