6-(Trifluoromethyl)nicotinic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20074G1 AS20074G25 AS20074G5
ACTIAS20074G1EA 0 GBP
ACTIAS20074G1 ACTIAS20074G25 ACTIAS20074G5
6-(Trifluoromethyl)nicotinic acid
6-(Trifluoromethyl)nicotinic acid
Formula: C₇H₄F₃NO₂
MDL Number: MFCD00792430
CAS Number: 231291-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR