2-Bromo-4'-(1-pyrrolidinyl)acetophenone

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(Pyrrolidin-1-yl)phenacyl bromide

OT-0423-5G OT-0423-25G OT-0423-1G OT-0423-100G
COBBOT-0423-5GEA 0 GBP
COBBOT-0423-5G COBBOT-0423-25G COBBOT-0423-1G COBBOT-0423-100G
2-Bromo-4'-(1-pyrrolidinyl)acetophenone
2-Bromo-4'-(1-pyrrolidinyl)acetophenone
Formula: C₁₂H₁₄BrNO
MDL Number: MFCD01075690
CAS Number: 216144-18-2
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR