Tetrahydropyran-4-carboxylic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS20984G25 AS20984G5
ACTIAS20984G25EA 0 GBP
ACTIAS20984G25 ACTIAS20984G5
Tetrahydropyran-4-carboxylic acid
Tetrahydropyran-4-carboxylic acid
Formula: C₆H₁₀O₃
MDL Number: MFCD00031016
CAS Number: 5337-03-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR