3-(1,3-Oxazol-5-yl)aniline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-(5-Oxazolyl)aniline

AS20650G1 AS20650G25 AS20650G5
ACTIAS20650G1EA 0 GBP
ACTIAS20650G1 ACTIAS20650G25 ACTIAS20650G5
3-(1,3-Oxazol-5-yl)aniline
3-(1,3-Oxazol-5-yl)aniline
Formula: C₉H₈N₂O
MW: 160.1700 g/mol
MDL Number: MFCD00052191
CAS Number: 157837-31-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR