3-Iodo-4-methylbenzotrifluoride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2652-5G HC-2652-1G
COBBHC-2652-5GEA 0 GBP
COBBHC-2652-5G COBBHC-2652-1G
3-Iodo-4-methylbenzotrifluoride
3-Iodo-4-methylbenzotrifluoride
Formula: C₈H₆F₃I
MDL Number: MFCD15143543
CAS Number: 13055-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR