9,10-Anthraquinone-2-carboxylic acid ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

H60929.03 H60929.06
H60929.03EA 41.9 GBP
H60929.03 H60929.06
9,10-Anthraquinone-2-carboxylic acid ≥98%
9,10-Anthraquinone-2-carboxylic acid
Formula: C₁₅H₈O₄
Melting Pt: 287…289 °C
Storage Temperature: Ambient
MDL Number: MFCD00001231
CAS Number: 117-78-2
EINECS: 204-207-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR