Portable balances, Core™ series
Supplier: ADAM EQUIPMENT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CQT1501 CQT2000 CQT202 CQT251 CQT2601 CQT5000 CQT601
611-2898EA 148 GBP
611-2898 611-2899 611-2900 611-2901 611-2902 611-2903 611-2904
Portable balances, Core™ series
Balances and Scales
The Core™ series balances from Adam Equipment provide simple operation and economy. Features low battery indicator, auto-off and auto-sleep function to save battery life.
  • Up to nine weighing units, full range tare
  • Backlit LCD and colour coded sealed keypad
  • External calibration
Units feature a lightweight, portable design, with overload protection, security locking point* and stainless steel pan. Applications include below balance weighing and weight accumulation.
Models with 120 mm pan are stackable for easy storage and are supplied with a removable draught shield.
Ordering information: * The optional security lock and cable are listed in the accessories below. Please contact Avantor for details on other accessories, including calibration weights.
Delivery information: Supplied with AC adapter and balance hanger. Balances can be battery-operated  (6 x AA type); batteries are not supplied.
Caution: Please note these products are not available in Republic of Ireland and Northern Ireland.