Cartridges, Dionex™, InGuard™
Supplier: DIONEX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
074034 074035 074036 074037 074038
DION074034EA 0 GBP
DION074034 DION074035 DION074036 DION074037 DION074038
Cartridges, Dionex™, InGuard™
Chromatography Columns
Dionex™ InGuard™ HRP cartridges are ideal for the removal of organic matrix material over a wide range of hydrophobicity including fats from whole milk.
  • Time-saving, in-line sample pretreatment
  • Simplified with automated workflow
  • Reproducible PPM-level determinations in concentrated matrices
  • Increased analytical column lifetimes
  • Leak-free, no-channelling cartridge design
The resin is water-wettable therefore, 100% aqueous samples can be pretreated without disruption of the column bed. Easy installation with standard 10-32 connections. Used singly or inMore Product Information