(2-Acetamidophenyl)boronic acid pinacol ester
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BB-2173-5G
BB-2173-1G
COBBBB-2173-5GEA
0
GBP
COBBBB-2173-5G
COBBBB-2173-1G
(2-Acetamidophenyl)boronic acid pinacol ester
(2-Acetamidophenyl)boronic acid pinacol ester
Formula:
C₁₄H₂₀BNO₃ |
MDL Number:
MFCD02179480 CAS Number: 380430-61-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...