2-Cyano-3-methylpyridine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS21747G100
AS21747G25
ACTIAS21747G100EA
0
GBP
ACTIAS21747G100
ACTIAS21747G25
2-Cyano-3-methylpyridine
2-Cyano-3-methylpyridine
Formula:
C₇H₆N₂ Boiling Pt: 139…142 °C (38 mmHg) Melting Pt: 87…90 °C |
MDL Number:
MFCD00190585 CAS Number: 20970-75-6 EINECS: 244-131-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...