2-Methyl-6-(tributylstannyl)pyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS51192G1 AS51192G5
ACTIAS51192G1EA 55 GBP
ACTIAS51192G1 ACTIAS51192G5
2-Methyl-6-(tributylstannyl)pyridine
2-Methyl-6-(tributylstannyl)pyridine
Formula: C₁₈H₃₃NSn
MW: 382.16 g/mol
MDL Number: MFCD07787401
CAS Number: 259807-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR