6-Chloro-3-(tributylstannyl)pyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS51354G1 AS51354G5
ACTIAS51354G1EA 0 GBP
ACTIAS51354G1 ACTIAS51354G5
6-Chloro-3-(tributylstannyl)pyridine
6-Chloro-3-(tributylstannyl)pyridine
Formula: C₁₇H₃₀ClNSn
MDL Number: MFCD01318969
CAS Number: 183545-05-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR