4-Chloro-2-(tributylstannyl)pyridine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
AS50398G5
AS50398
AS50398G1
ACTIAS50398G5EA
0
GBP
ACTIAS50398G5
ACTIAS50398
ACTIAS50398G1
4-Chloro-2-(tributylstannyl)pyridine
4-Chloro-2-(tributylstannyl)pyridine
Formula:
C₁₇H₃₀ClNSn |
MDL Number:
MFCD09870067 CAS Number: 1204580-71-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...