2,6-Dichloro-4-methylpyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS50949G1 AS50949G25 AS50949G5
ACTIAS50949G1EA 14 GBP
ACTIAS50949G1 ACTIAS50949G25 ACTIAS50949G5
2,6-Dichloro-4-methylpyridine
2,6-Dichloro-4-methylpyridine
Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
MDL Number: MFCD09264302
CAS Number: 39621-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR