2,6-Dichloro-3-methylpyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7271-1G PY-7271-5G
COBBPY-7271-1GEA 0 GBP
COBBPY-7271-1G COBBPY-7271-5G
2,6-Dichloro-3-methylpyridine
2,6-Dichloro-3-methylpyridine
Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
MDL Number: MFCD00137847
CAS Number: 58584-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR